Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256169.1 | NP_001243098.1 | 3 | 359 | |
NM_019101.2 | NP_061974.2 | 3 | 404 | ||
NR_045828.1 | 3 | 366 | |||
XM_006715150.3 | XP_006715213.1 | 3 | 388 | ||
GenBank mRNA | AF118393.3 | - | 3 | 364 | |
AF161454.1 | - | 3 | 325 | ||
AJ245434.1 | - | 3 | 380 | ||
BC020683.1 | - | 3 | 383 | 4733993 | |
BF344944.1 | - | 3 | 340 | ||
BG564719.1 | - | 3 | 405 | ||
CB054016.1 | - | 3 | 366 | ||
CN428415.1 | - | 3 | 346 |