Gene Transcripts

Gene Symbol
CMC2
Entrez Gene ID
56942
Gene Name
C-X9-C motif containing 2
Gene Aliases
2310061C15Rik, C16orf61, DC13
Chromosome Location
Chr. 16: 80975800 - 81007046 on Build GRCh38
UniGene ID
Hs.55028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020188.3 NP_064573.1 4 395
XM_017023461.1 XP_016878950.1 5 583
XM_017023462.1 XP_016878951.1 6 708
XM_017023464.1 XP_016878953.1 3 452
XM_017023465.1 XP_016878954.1 4 571
XM_017023466.1 XP_016878955.1 2 356
XM_017023467.1 XP_016878956.1 6 478
XM_017023468.1 XP_016878957.1 5 388
XM_017023469.1 XP_016878958.1 5 462
XM_017023470.1 XP_016878959.1 5 386
GenBank mRNA AF201935.1 - 4 371
AK123193.1 - 3 1750
BC032631.1 - 4 399 5399193

View Full Product Details