Gene Transcripts

Gene Symbol
NFE4
Entrez Gene ID
58160
Gene Name
nuclear factor, erythroid 4
Gene Aliases
NF-E4
Chromosome Location
Chr. 7: 102973437 - 102978667 on Build GRCh38
UniGene ID
Hs.621233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085386.1 NP_001078855.1 2 609
GenBank mRNA AY258907.1 - 2 609

View Full Product Details