Gene Transcripts
- Gene Symbol
- UBL5
- Entrez Gene ID
- 59286
- Gene Name
- ubiquitin like 5
- Gene Aliases
- HUB1
- Chromosome Location
- Chr. 19: 9827880 - 9830121 on Build GRCh38
- UniGene ID
- Hs.534477
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001048241.2 |
NP_001041706.1
|
3, 4 |
260 |
|
NM_024292.3 |
NP_077268.1
|
3, 4 |
328 |
|
GenBank mRNA |
AF313915.1 |
- |
3, 4 |
209 |
|
AK128885.1 |
- |
3, 4 |
328 |
|
AK311889.1 |
- |
3, 4 |
248 |
|
AV682189.1 |
- |
3, 4 |
260 |
|
BC007053.1 |
- |
3, 4 |
282 |
3827384 |
BC110839.1 |
- |
3, 4 |
176 |
6474495 |
BP213482.1 |
- |
3, 4 |
248 |
|
BT007355.1 |
- |
2, 3 |
144 |
|
View Full Product Details