Gene Transcripts

Gene Symbol
ATG101
Entrez Gene ID
60673
Gene Name
autophagy related 101
Gene Aliases
C12orf44
Chromosome Location
Chr. 12: 52069974 - 52077495 on Build GRCh38
UniGene ID
Hs.9911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098673.1 NP_001092143.1 3 485
NM_021934.4 NP_068753.2 3 656
GenBank mRNA AF218031.1 - 3 597
AK021835.1 - 3 654
BC005151.2 - 3 608 3508173
BC009937.2 - 3 590 4303452
BP309508.1 - 3 456

View Full Product Details