Gene Transcripts

Gene Symbol
EXO5
Entrez Gene ID
64789
Gene Name
exonuclease 5
Gene Aliases
C1orf176, DEM1, Exo V, hExo5
Chromosome Location
Chr. 1: 40508719 - 40516556 on Build GRCh38
UniGene ID
Hs.59584
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022774.1 NP_073611.1 3 569
XM_005271125.3 XP_005271182.1 4 817
XM_005271126.3 XP_005271183.1 3 629
XM_005271128.3 XP_005271185.1 2 566
XM_011541970.2 XP_011540272.1 2 511
XM_017002099.1 XP_016857588.1 4 1241
XM_017002100.1 XP_016857589.1 4 796
XM_017002101.1 XP_016857590.1 4 767
XM_017002102.1 XP_016857591.1 3 1169
XM_017002103.1 XP_016857592.1 3 595
XM_017002104.1 XP_016857593.1 2 548
GenBank mRNA AK023245.1 - 2 1396
AK024797.1 - 3 569
AK094092.1 - 3 1391
BC021969.2 - 4 730 3952573

View Full Product Details