Gene Transcripts
- Gene Symbol
- USP46
- Entrez Gene ID
- 64854
- Gene Name
- ubiquitin specific peptidase 46
- Gene Aliases
- Chromosome Location
- Chr. 4: 52590960 - 52659335 on Build GRCh38
- UniGene ID
- Hs.7966
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134223.1 |
NP_001127695.1
|
4, 5 |
824 |
|
NM_001286767.1 |
NP_001273696.1
|
4, 5 |
712 |
|
NM_001286768.1 |
NP_001273697.1
|
5, 6 |
856 |
|
NM_022832.3 |
NP_073743.2
|
4, 5 |
748 |
|
XM_017008557.1 |
XP_016864046.1
|
4, 5 |
714 |
|
GenBank mRNA |
AK022614.1 |
- |
4, 5 |
568 |
|
AK024318.1 |
- |
4, 5 |
689 |
|
AK296493.1 |
- |
5, 6 |
829 |
|
AK299883.1 |
- |
4, 5 |
625 |
|
AK302438.1 |
- |
4, 5 |
823 |
|
BC037574.1 |
- |
4, 5 |
697 |
5529667 |
GU455414.1 |
- |
4, 5 |
563 |
|
View Full Product Details