Gene Transcripts

Gene Symbol
INTS3
Entrez Gene ID
65123
Gene Name
integrator complex subunit 3
Gene Aliases
C1orf193, C1orf60, INT3, SOSS-A, SOSSA
Chromosome Location
Chr. 1: 153728050 - 153774808 on Build GRCh38
UniGene ID
Hs.438723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324475.1 NP_001311404.1 24 2584
NM_023015.4 NP_075391.3 23 2925
XM_005245459.2 XP_005245516.1 23 2942
XM_005245461.2 XP_005245518.1 17 1615
XM_006711490.3 XP_006711553.1 24 2395
XM_011509906.2 XP_011508208.1 23 2307
XM_017002132.1 XP_016857621.1 13 1183
GenBank mRNA AK025572.1 - 9 746
AK055174.1 - 3 1110
AK074979.1 - 16 1623
AK126799.1 - 3 1463
AK225294.1 - 9 746
AK290025.1 - 24 2578
AK298746.1 - 11 1075
AK304874.1 - 17 2014
AL080172.1 - 18 1824
AL832133.1 - 22 3960
BC025254.1 - 7 585
BC054513.1 - 10 961
BC073985.1 - 7 597
BC098431.1 - 9 785
BC105092.1 - 23 2388 8144095
BC105094.1 - 23 2388 8144097
BC116458.1 - 23 2920 5752962
BX640786.1 - 23 2911
BX640950.1 - 23 2926
CR749212.1 - 21 4452
CR749376.1 - 22 4577

View Full Product Details