Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024949.5 | NP_079225.5 | 7, 8 | 1064 | |
XM_011532269.2 | XP_011530571.1 | 7, 8 | 1697 | ||
GenBank mRNA | AK126057.1 | - | 6, 7 | 729 | |
BM046889.1 | - | 500 | |||
BX647378.1 | - | 5, 6 | 655 |