Gene Transcripts

Gene Symbol
SLC25A28
Entrez Gene ID
81894
Gene Name
solute carrier family 25 member 28
Gene Aliases
MFRN2, MRS3/4, MRS4L, NPD016
Chromosome Location
Chr. 10: 99610518 - 99659329 on Build GRCh38
UniGene ID
Hs.403790
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031212.3 NP_112489.3 4 903
XM_005270208.1 XP_005270265.1 4 893
XM_005270211.4 XP_005270268.1 4 507
XM_006718005.3 XP_006718068.1 4 567
XM_011540239.2 XP_011538541.1 4 789
XM_011540240.1 XP_011538542.1 5 2039
XM_017016741.1 XP_016872230.1 4 1975
XM_017016742.1 XP_016872231.1 4 1796
GenBank mRNA AF267854.1 - 1 590
AF327402.1 - 4 803
AF327403.1 - 4 1244
AJ303077.1 - 4 803
AJ303078.1 - 4 1244
AK056782.1 - 1 943
AL831943.1 - 1 465
BC047312.1 - 4 850
BC058937.1 - 4 812 6139015
BC076399.1 - 4 843 5728224
BC094821.1 - 4 586

View Full Product Details