Gene Transcripts

Gene Symbol
OBSCN
Entrez Gene ID
84033
Gene Name
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases
ARHGEF30, UNC89
Chromosome Location
Chr. 1: 228208013 - 228381431 on Build GRCh38
UniGene ID
Hs.650039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098623.2 NP_001092093.2 56 15586
NM_001271223.2 NP_001258152.2 67 18457
NM_052843.3 NP_443075.3 56 15586
XM_005273287.4 XP_005273344.1 68 18837
XM_005273291.4 XP_005273348.1 68 18801
XM_005273298.4 XP_005273355.1 67 18528
XM_005273307.4 XP_005273364.1 63 17715
XM_006711822.3 XP_006711885.1 66 18261
XM_006711823.3 XP_006711886.1 66 18261
XM_006711827.3 XP_006711890.1 60 16676
XM_006711829.3 XP_006711892.1 59 16400
XM_011544297.2 XP_011542599.1 60 16677
XM_011544299.2 XP_011542601.1 59 16401
XM_017002443.1 XP_016857932.1 68 18882
XM_017002444.1 XP_016857933.1 68 18845
XM_017002445.1 XP_016857934.1 68 18882
XM_017002446.1 XP_016857935.1 68 18882
XM_017002447.1 XP_016857936.1 68 18879
XM_017002448.1 XP_016857937.1 68 18732
XM_017002449.1 XP_016857938.1 67 18619
XM_017002450.1 XP_016857939.1 67 18619
XM_017002451.1 XP_016857940.1 67 18619
XM_017002452.1 XP_016857941.1 67 18619
XM_017002453.1 XP_016857942.1 67 18606
XM_017002454.1 XP_016857943.1 67 18606
XM_017002455.1 XP_016857944.1 67 18606
XM_017002456.1 XP_016857945.1 67 18606
XM_017002457.1 XP_016857946.1 67 18606
XM_017002458.1 XP_016857947.1 66 18355
XM_017002459.1 XP_016857948.1 66 18355
XM_017002460.1 XP_016857949.1 66 18355
XM_017002461.1 XP_016857950.1 66 18342
XM_017002462.1 XP_016857951.1 66 18330
XM_017002463.1 XP_016857952.1 66 18309
XM_017002464.1 XP_016857953.1 66 18258
XM_017002465.1 XP_016857954.1 65 18091
XM_017002466.1 XP_016857955.1 65 18091
XM_017002467.1 XP_016857956.1 65 18091
XM_017002468.1 XP_016857957.1 64 17827
XM_017002469.1 XP_016857958.1 62 17299
XM_017002470.1 XP_016857959.1 57 15980
XM_017002471.1 XP_016857960.1 68 18881
XM_017002472.1 XP_016857961.1 68 18881
XM_017002473.1 XP_016857962.1 68 18881
GenBank mRNA AJ002535.1 - 57 15583

View Full Product Details