Gene Transcripts

Gene Symbol
TRIM55
Entrez Gene ID
84675
Gene Name
tripartite motif containing 55
Gene Aliases
MURF-2, RNF29, muRF2
Chromosome Location
Chr. 8: 66112667 - 66175485 on Build GRCh38
UniGene ID
Hs.85524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033058.2 NP_149047.2 2 479
NM_184085.1 NP_908973.1 2 479
NM_184086.1 NP_908974.1 2 479
NM_184087.1 NP_908975.1 2 479
XM_005251316.4 XP_005251373.1 4 1504
XM_011517613.2 XP_011515915.1 4 1504
XM_011517614.2 XP_011515916.1 4 1427
XM_011517615.2 XP_011515917.1 4 1504
XM_011517616.2 XP_011515918.1 4 1504
XM_017013908.1 XP_016869397.1 2 197
GenBank mRNA AJ243488.1 - 2 479
AJ243489.1 - 2 479
AJ277493.1 - 2 479
AJ291712.1 - 2 385
AJ431704.1 - 2 479
AK091310.1 - 2 629
AK091728.1 - 2 299
BC007750.2 - 2 385 4110783
BT007212.1 - 2 205

View Full Product Details