Gene Transcripts

Gene Symbol
MFSD2A
Entrez Gene ID
84879
Gene Name
major facilitator superfamily domain containing 2A
Gene Aliases
MCPH15, MFSD2, NLS1
Chromosome Location
Chr. 1: 39955112 - 39969968 on Build GRCh38
UniGene ID
Hs.655177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136493.2 NP_001129965.1 11 1371
NM_001287808.1 NP_001274737.1 9 1072
NM_001287809.1 NP_001274738.1 11 1215
NM_032793.4 NP_116182.2 11 1332
XM_005271285.1 XP_005271342.1 11 1296
XM_011542312.2 XP_011540614.1 10 1215
GenBank mRNA AB073383.1 - 11 1296
AF289609.1 - 11 1288
AF370364.1 - 6 1592
AK074186.1 - 11 1237
AK075183.1 - 11 1299
AK093223.1 - 11 1333
AK291540.1 - 11 1294
AK297991.1 - 11 1179
AK301533.1 - 9 1039
AY358636.1 - 11 1314
BC006353.2 - 7 667
BC011587.1 - 11 1306 4646430
BC092414.1 - 11 1274 30387073

View Full Product Details