Gene Transcripts

Gene Symbol
NKD2
Entrez Gene ID
85409
Gene Name
naked cuticle homolog 2
Gene Aliases
Naked2
Chromosome Location
Chr. 5: 1008845 - 1038812 on Build GRCh38
UniGene ID
Hs.240951
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271082.1 NP_001258011.1 7 541
NM_033120.3 NP_149111.1 7 541
XM_005248382.3 XP_005248439.1 7 658
XM_005248383.3 XP_005248440.1 7 658
XM_011514150.2 XP_011512452.1 7 658
XM_017010000.1 XP_016865489.1 2 844
GenBank mRNA AB062887.1 - 7 450
AF358137.1 - 7 462
BC004940.1 - 2 69
BC012176.1 - 7 541 4649935

View Full Product Details