Gene Transcripts

Gene Symbol
MLIP
Entrez Gene ID
90523
Gene Name
muscular LMNA-interacting protein
Gene Aliases
C6orf142
Chromosome Location
Chr. 6: 54018916 - 54266325 on Build GRCh38
UniGene ID
Hs.449276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281746.1 NP_001268675.1 3 426
NM_001281747.1 NP_001268676.1 3 408
NM_138569.2 NP_612636.2 3 426
XM_005249476.4 XP_005249533.1 3 379
XM_005249478.4 XP_005249535.1 2 233
XM_005249480.4 XP_005249537.1 3 379
XM_006715245.3 XP_006715308.2 3 518
XM_017011465.1 XP_016866954.1 3 515
XM_017011466.1 XP_016866955.1 3 512
XM_017011467.1 XP_016866956.1 3 521
XM_017011468.1 XP_016866957.1 3 520
XM_017011469.1 XP_016866958.1 3 312
XM_017011470.1 XP_016866959.1 3 610
GenBank mRNA AK055530.1 - 3 351
AK294797.1 - 2 223
AK294890.1 - 3 379
AK311190.1 - 3 379
AK316157.1 - 3 359
BC009010.1 - 2 173 4158300
BX091237.1 - 3 293
BX647567.1 - 3 372
DA761851.1 - 3 426
DC322019.1 - 2 252
KU178822.1 - 2 127
KU178823.1 - 3 283
KU178824.1 - 3 283
KU178825.1 - 3 283
KU178826.1 - 3 283

View Full Product Details