Gene Transcripts

Gene Symbol
LONRF1
Entrez Gene ID
91694
Gene Name
LON peptidase N-terminal domain and ring finger 1
Gene Aliases
RNF191
Chromosome Location
Chr. 8: 12721897 - 12755485 on Build GRCh38
UniGene ID
Hs.180178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152271.3 NP_689484.3 12 2295
XM_005273685.3 XP_005273742.1 12 2264
XM_011544694.2 XP_011542996.1 12 2047
GenBank mRNA AK074329.1 - 7 847
AK297266.1 - 11 1432
AK300538.1 - 8 1041
BC004538.2 - 4 431
BC106042.1 - 12 1561
BC117381.1 - 9 1226 40125932
BC117385.1 - 9 1226 40125936

View Full Product Details