Gene Transcripts
- Gene Symbol
- UBE2Q2
- Entrez Gene ID
- 92912
- Gene Name
- ubiquitin conjugating enzyme E2 Q2
- Gene Aliases
- Chromosome Location
- Chr. 15: 75843286 - 75901047 on Build GRCh38
- UniGene ID
- Hs.23033
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145335.1 |
NP_001138807.1
|
11, 12 |
1299 |
|
NM_001284382.1 |
NP_001271311.1
|
10, 11 |
1313 |
|
NM_173469.3 |
NP_775740.1
|
11, 12 |
1418 |
|
XM_005254788.1 |
XP_005254845.1
|
8, 9 |
953 |
|
XM_017022728.1 |
XP_016878217.1
|
7, 8 |
876 |
|
GenBank mRNA |
AK000617.1 |
- |
10, 11 |
1292 |
|
AK294199.1 |
- |
11, 12 |
959 |
|
AK296234.1 |
- |
11, 12 |
1299 |
|
AL832429.1 |
- |
11, 12 |
1243 |
|
BC017708.1 |
- |
11, 12 |
1245 |
3907760 |
BC034342.1 |
- |
3, 4 |
211 |
|
View Full Product Details