Gene Transcripts

Gene Symbol
SFXN5
Entrez Gene ID
94097
Gene Name
sideroflexin 5
Gene Aliases
BBG-TCC
Chromosome Location
Chr. 2: 72942036 - 73072234 on Build GRCh38
UniGene ID
Hs.368171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144579.2 NP_653180.1 13, 14 1072
XM_005264648.1 XP_005264705.1 13, 14 955
XM_006712147.1 XP_006712210.1 11, 12 776
XM_011533187.2 XP_011531489.1 14, 15 2617
XM_011533195.2 XP_011531497.1 12, 13 2418
XM_017005340.1 XP_016860829.1 13, 14 2472
XM_017005343.1 XP_016860832.1 12, 13 1557
XM_017005344.1 XP_016860833.1 13, 14 1704
GenBank mRNA AK289731.1 - 13, 14 961
AK294751.1 - 13, 14 1009
AK295637.1 - 13, 14 860
AK309091.1 - 4, 5 542
AK314770.1 - 11, 12 758
BC101311.2 - 13, 14 1072 40024811
BC101312.1 - 13, 14 1072 40024812
BC101313.2 - 13, 14 1072 40024813

View Full Product Details