Gene Transcripts

Gene Symbol
C1QTNF1
Entrez Gene ID
114897
Gene Name
C1q and tumor necrosis factor related protein 1
Gene Aliases
CTRP1, GIP, ZSIG37
Chromosome Location
Chr. 17: 79022934 - 79049788 on Build GRCh38
UniGene ID
Hs.201398
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030968.3 NP_112230.1 4 977
NM_153372.1 NP_699203.1 4 713
NM_198593.3 NP_940995.1 4 793
NM_198594.2 NP_940996.1 3 808
NR_049769.1 4 831
XM_006721663.1 XP_006721726.1 3 838
XM_006721664.1 XP_006721727.1 6 1425
XM_006721666.1 XP_006721729.1 5 1041
XM_017024126.1 XP_016879615.1 3 1082
GenBank mRNA AF232905.1 - 3 920
AF329840.1 - 4 714
AJ272138.1 - 3 920
AK055541.1 - 3 623
AK075175.1 - 2 318
AK172767.1 - 4 814
AK292034.1 - 4 831
AY358424.1 - 4 943
BC021553.1 - 4 599 4622952

View Full Product Details