Gene Transcripts

Gene Symbol
LRG1
Entrez Gene ID
116844
Gene Name
leucine rich alpha-2-glycoprotein 1
Gene Aliases
HMFT1766, LRG
Chromosome Location
Chr. 19: 4537215 - 4540024 on Build GRCh38
UniGene ID
Hs.655559
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052972.2 NP_443204.1 2 934
GenBank mRNA AB073607.1 - 2 946
AF403428.1 - 2 934
AK027382.1 - 2 946
AK303861.1 - 3 895
AK314506.1 - 2 946
BC034389.1 - 2 935 5187278
BC070198.1 - 2 991 4692618

View Full Product Details