Gene Transcripts

Gene Symbol
SLAIN1
Entrez Gene ID
122060
Gene Name
SLAIN motif family member 1
Gene Aliases
C13orf32
Chromosome Location
Chr. 13: 77697854 - 77764242 on Build GRCh38
UniGene ID
Hs.349955
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040153.3 NP_001035243.2 6 1088
NM_001242868.1 NP_001229797.1 6 1597
NM_001242869.1 NP_001229798.1 5 860
NM_001242870.1 NP_001229799.1 4 710
NM_001242871.1 NP_001229800.1 5 1136
NM_144595.3 NP_653196.1 5 1052
XM_011534929.2 XP_011533231.1 7 1477
XM_011534930.1 XP_011533232.1 6 990
GenBank mRNA AK054608.1 - 5 832
AK094337.1 - 1 633
AK289600.1 - 5 687
AK289714.1 - 5 842
AK294201.1 - 6 1088
AK295423.1 - 5 912
AL834203.1 - 4 574
AL834332.1 - 5 845
BC045177.1 - 5 861
BC111497.1 - 5 843 6148614

View Full Product Details