Gene Transcripts
- Gene Symbol
- NKAIN4
- Entrez Gene ID
- 128414
- Gene Name
- Na+/K+ transporting ATPase interacting 4
- Gene Aliases
- C20orf58, FAM77A, bA261N11.2
- Chromosome Location
- Chr. 20: 63240784 - 63254904 on Build GRCh38
- UniGene ID
- Hs.46627
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152864.3 |
NP_690603.3
|
3, 4 |
361 |
|
XM_005260192.1 |
XP_005260249.1
|
3, 4 |
323 |
|
XM_005260194.3 |
XP_005260251.1
|
3, 4 |
782 |
|
XM_011528527.1 |
XP_011526829.1
|
3, 4 |
323 |
|
XM_011528528.2 |
XP_011526830.1
|
3, 4 |
785 |
|
XM_011528529.2 |
XP_011526831.1
|
3, 4 |
771 |
|
XM_017027636.1 |
XP_016883125.1
|
3, 4 |
293 |
|
XM_017027637.1 |
XP_016883126.1
|
3, 4 |
775 |
|
XM_017027638.1 |
XP_016883127.1
|
3, 4 |
781 |
|
XM_017027639.1 |
XP_016883128.1
|
3, 4 |
767 |
|
GenBank mRNA |
AK225765.1 |
- |
3, 4 |
315 |
|
BC041812.1 |
- |
3, 4 |
359 |
5302801 |
BG708360.1 |
- |
3, 4 |
365 |
|
View Full Product Details