Gene Transcripts

Gene Symbol
MIR1-1HG
Entrez Gene ID
128826
Gene Name
MIR1-1 host gene
Gene Aliases
C20orf166, MIR133A2HG, dJ353C17.1
Chromosome Location
Chr. 20: 62550453 - 62570764 on Build GRCh38
UniGene ID
Hs.86507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302812.1 NP_001289741.1 5 543
NM_178463.3 NP_848558.1 4 924
GenBank mRNA AK056985.1 - 4 940
BC110595.2 - 4 599 40022808
DA896197.1 - 5 540

View Full Product Details