Gene Transcripts

Gene Symbol
VCX
Entrez Gene ID
26609
Gene Name
variable charge, X-linked
Gene Aliases
VCX-10r, VCX-B1, VCX1, VCX10R, VCXB1
Chromosome Location
Chr. X: 7842262 - 7844143 on Build GRCh38
UniGene ID
Hs.278906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013452.2 NP_038480.2 1 28
GenBank mRNA AF167081.2 - 1 28

View Full Product Details