Gene Transcripts

Gene Symbol
PABPC1L2A
Entrez Gene ID
340529
Gene Name
poly(A) binding protein cytoplasmic 1 like 2A
Gene Aliases
RBM32A
Chromosome Location
Chr. X: 73077338 - 73079512 on Build GRCh38
UniGene ID
Hs.450320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012977.2 NP_001012995.1 1 2064
GenBank mRNA AY730282.1 - 1 463

View Full Product Details