Gene Transcripts

Gene Symbol
TDRP
Entrez Gene ID
157695
Gene Name
testis development related protein
Gene Aliases
C8orf42, Inm01, TDRP1, TDRP2
Chromosome Location
Chr. 8: 489790 - 545781 on Build GRCh38
UniGene ID
Hs.289293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256113.1 NP_001243042.1 2, 3 296
NM_175075.4 NP_778250.2 3, 4 464
XM_006716232.3 XP_006716295.1 2, 3 444
XM_017013113.1 XP_016868602.1 2, 3 778
GenBank mRNA AY194292.2 - 2, 3 282
BC136248.1 - 2, 3 443 9020235
BC136249.1 - 2, 3 443 9020236
BX648606.1 - 1, 2 67
CN430635.1 - 2, 3 275
DB461052.2 - 3, 4 458
FJ381685.1 - 2, 3 288

View Full Product Details