Gene Transcripts
- Gene Symbol
- CSAG1
- Entrez Gene ID
- 158511
- Gene Name
- chondrosarcoma associated gene 1
- Gene Aliases
- CSAGE, CT24.1
- Chromosome Location
- Chr. X: 152727484 - 152733736 on Build GRCh38
- UniGene ID
- Hs.423718
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BF570999.1 |
- |
4 |
423 |
|
BM810082.1 |
- |
4 |
405 |
|
View Full Product Details