Gene Transcripts

Gene Symbol
MAJIN
Entrez Gene ID
283129
Gene Name
membrane anchored junction protein
Gene Aliases
C11orf85
Chromosome Location
Chr. 11: 64938230 - 64972104 on Build GRCh38
UniGene ID
Hs.567793
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037225.2 NP_001032302.1 6 592
NM_001318808.1 NP_001305737.1 6 1306
XM_005273918.2 XP_005273975.1 6 586
XM_006718502.2 XP_006718565.1 6 586
XM_006718503.3 XP_006718566.1 6 472
XM_006718504.2 XP_006718567.1 6 586
XM_011544940.2 XP_011543242.1 6 467
XM_011544941.2 XP_011543243.1 1 317
GenBank mRNA AK093130.1 - 6 1300
BC106951.2 - 5 387 40035116
BC106952.1 - 5 387 40035117
DB445650.2 - 6 592
LC068587.1 - 4 313

View Full Product Details