Gene Transcripts

Gene Symbol
PPIL6
Entrez Gene ID
285755
Gene Name
peptidylprolyl isomerase like 6
Gene Aliases
PPIase, RSPH12, bA425D10.6, dJ919F19.1
Chromosome Location
Chr. 6: 109390215 - 109441171 on Build GRCh38
UniGene ID
Hs.32234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111298.2 NP_001104768.2 8 1454
NM_001286360.1 NP_001273289.1 6 1280
NM_173672.4 NP_775943.1 7 1376
NR_104429.1 8 1496
XM_011535765.2 XP_011534067.1 7 818
GenBank mRNA AK293674.1 - 8 948
AK315790.1 - 7 849
BC038716.1 - 7 835 5268756
DQ363562.1 - 7 1376
DQ423529.1 - 8 1454

View Full Product Details