Gene Transcripts

Gene Symbol
SLC35F4
Entrez Gene ID
341880
Gene Name
solute carrier family 35 member F4
Gene Aliases
C14orf36, c14_5373
Chromosome Location
Chr. 14: 57563922 - 57982329 on Build GRCh38
UniGene ID
Hs.28280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206920.1 NP_001193849.1 2 259
NM_001306087.1 NP_001293016.1 2 259
XM_011536720.2 XP_011535022.1 4 4038
XM_011536721.2 XP_011535023.1 4 4038
XM_011536723.2 XP_011535025.1 2 524
XM_011536724.2 XP_011535026.1 2 1129
XM_011536725.1 XP_011535027.1 3 451
XM_017021258.1 XP_016876747.1 3 444
XM_017021259.1 XP_016876748.1 4 4038
XM_017021260.1 XP_016876749.1 3 451
XM_017021261.1 XP_016876750.1 2 330
GenBank mRNA BC101318.1 - 2 200
BC101319.1 - 2 1270
BC101320.1 - 2 206
BC101321.1 - 2 206

View Full Product Details