Gene Transcripts

Gene Symbol
LCE1B
Entrez Gene ID
353132
Gene Name
late cornified envelope 1B
Gene Aliases
LEP2, SPRL2A
Chromosome Location
Chr. 1: 152811971 - 152813109 on Build GRCh38
UniGene ID
Hs.375103
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178349.1 NP_848126.1 1 1086
GenBank mRNA BC031811.1 - 1 1086

View Full Product Details