Gene Transcripts

Gene Symbol
OR4X1
Entrez Gene ID
390113
Gene Name
olfactory receptor family 4 subfamily X member 1 (gene/pseudogene)
Gene Aliases
OR11-104
Chromosome Location
Chr. 11: 48263861 - 48264778 on Build GRCh38
UniGene ID
Hs.553742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004726.1 NP_001004726.1 1 759

View Full Product Details