Gene Transcripts

Gene Symbol
OR5AK2
Entrez Gene ID
390181
Gene Name
olfactory receptor family 5 subfamily AK member 2
Gene Aliases
Chromosome Location
Chr. 11: 56988914 - 56989843 on Build GRCh38
UniGene ID
Hs.554520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005323.1 NP_001005323.1 1 489
GenBank mRNA BC137198.1 - 1 531

View Full Product Details