Gene Transcripts

Gene Symbol
EFCAB13
Entrez Gene ID
124989
Gene Name
EF-hand calcium binding domain 13
Gene Aliases
C17orf57
Chromosome Location
Chr. 17: 47323961 - 47441312 on Build GRCh38
UniGene ID
Hs.463303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195192.1 NP_001182121.1 10 1370
NM_152347.4 NP_689560.3 12 1658
GenBank mRNA AK097661.1 - 9 1217
BC036407.2 - 12 1638 5266195
BC037876.1 - 10 1310 5273577

View Full Product Details