Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021083.2 | NP_066569.1 | 2 | 467 | |
XM_011543978.2 | XP_011542280.1 | 2 | 581 | ||
GenBank mRNA | AY534238.1 | - | 2 | 591 | |
BC036019.1 | - | 2 | 508 | 5272840 | |
Z32684.2 | - | 2 | 467 |