Gene Transcripts
- Gene Symbol
- CTSA
- Entrez Gene ID
- 5476
- Gene Name
- cathepsin A
- Gene Aliases
- GLB2, GSL, NGBE, PPCA, PPGB
- Chromosome Location
- Chr. 20: 45890144 - 45898820 on Build GRCh38
- UniGene ID
- Hs.609336
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000308.3 |
NP_000299.2
|
8, 9 |
2012 |
|
NM_001127695.2 |
NP_001121167.1
|
8, 9 |
2058 |
|
NM_001167594.2 |
NP_001161066.1
|
7, 8 |
1961 |
|
NR_133656.1 |
|
8, 9 |
2019 |
|
GenBank mRNA |
AB209705.2 |
- |
8, 9 |
828 |
|
AK097786.1 |
- |
7, 8 |
725 |
|
AK172808.1 |
- |
8, 9 |
2016 |
|
AK304536.1 |
- |
7, 8 |
789 |
|
AK307701.1 |
- |
8, 9 |
805 |
|
AK312898.1 |
- |
8, 9 |
821 |
|
BC000597.2 |
- |
8, 9 |
814 |
3346150 |
BC093009.1 |
- |
8, 9 |
821 |
30348898 |
M22960.1 |
- |
7, 8 |
782 |
|
View Full Product Details