Gene Transcripts

Gene Symbol
PEX11B
Entrez Gene ID
8799
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11-BETA, PEX14B
Chromosome Location
Chr. 1: 145911348 - 145918924 on Build GRCh38
UniGene ID
Hs.714608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184795.1 NP_001171724.1 4 910
NM_003846.2 NP_003837.1 4 1104
NR_073491.1 4 1100
NR_073492.1 4 1094
NR_073493.2 4 1316
GenBank mRNA AB018080.1 - 4 886
AF093670.1 - 4 884
AK023991.1 - 4 868
AK094173.1 - 1 1704
AK301983.1 - 4 870
BC011963.1 - 4 884 3866690
BG435901.1 - 1 59

View Full Product Details