Gene Transcripts
- Gene Symbol
- MSRB3
- Entrez Gene ID
- 253827
- Gene Name
- methionine sulfoxide reductase B3
- Gene Aliases
- DFNB74
- Chromosome Location
- Chr. 12: 65278643 - 65466907 on Build GRCh38
- UniGene ID
- Hs.339024
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031679.2 |
NP_001026849.1
|
3, 4 |
458 |
|
NM_001193460.1 |
NP_001180389.1
|
4, 5 |
622 |
|
NM_001193461.1 |
NP_001180390.1
|
3, 4 |
320 |
|
NM_198080.3 |
NP_932346.1
|
2, 3 |
331 |
|
GenBank mRNA |
AK293084.1 |
- |
3, 4 |
428 |
|
AK299065.1 |
- |
2, 3 |
270 |
|
AK316365.1 |
- |
2, 3 |
302 |
|
AL833622.1 |
- |
3, 4 |
376 |
|
AY358229.1 |
- |
2, 3 |
266 |
|
BC040053.1 |
- |
4, 5 |
523 |
6025598 |
BX500552.1 |
- |
3, 4 |
322 |
|
BX648776.1 |
- |
3, 4 |
377 |
|
DC357690.1 |
- |
2, 3 |
331 |
|
View Full Product Details