Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130167.1 | NP_001123639.1 | 4 | 869 | |
NM_001130168.1 | NP_001123640.1 | 3 | 503 | ||
NM_182707.2 | NP_874366.1 | 4 | 869 | ||
XM_011526987.2 | XP_011525289.1 | 3 | 661 | ||
XM_011526988.2 | XP_011525290.1 | 4 | 852 | ||
GenBank mRNA | AK092458.1 | - | 1 | 920 | |
AK300230.1 | - | 3 | 502 | ||
AL545004.3 | - | 3 | 486 | ||
BC137500.1 | - | 4 | 869 | 9021500 | |
BC142628.1 | - | 4 | 857 | 40147510 | |
CR749812.1 | - | 4 | 868 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001184825.1 | NP_001171754.1 | 4 | 909 | |
NM_001184826.1 | NP_001171755.1 | 4 | 909 | ||
NM_001297773.1 | NP_001284702.1 | 4 | 857 | ||
NM_006905.2 | NP_008836.2 | 4 | 909 | ||
XM_005259065.3 | XP_005259122.1 | 3 | 629 | ||
XM_017026994.1 | XP_016882483.1 | 3 | 629 | ||
GenBank mRNA | AK056754.1 | - | 1 | 1674 | |
AK300152.1 | - | 4 | 572 | ||
AK300206.1 | - | 4 | 518 | ||
AK311491.1 | - | 4 | 810 | ||
BC022338.1 | - | 4 | 909 | 4732357 | |
BC058285.1 | - | 4 | 833 | 4770394 | |
CR450329.1 | - | 4 | 771 | ||
J04539.1 | - | 4 | 831 | ||
M17908.1 | - | 4 | 837 | ||
M20879.1 | - | 4 | 857 | ||
M20881.1 | - | 4 | 888 | ||
M21822.1 | - | 4 | 844 | ||
M25385.1 | - | 4 | 807 | ||
M33663.1 | - | 4 | 834 | ||
M33664.1 | - | 4 | 771 | ||
M34715.1 | - | 4 | 932 | ||
M37397.1 | - | 4 | 861 |