Gene Transcripts

Gene Symbol
UQCRH
Entrez Gene ID
7388
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Gene Aliases
QCR6, UQCR8
Chromosome Location
Chr. 1: 46303613 - 46316777 on Build GRCh38
UniGene ID
Hs.481571
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297565.1 NP_001284494.1 5 679
NM_001297566.1 NP_001284495.1 3 504
NM_006004.3 NP_005995.2 4 531
GenBank mRNA BC001426.2 - 4 431
BC001934.1 - 4 436 2823237
BC015177.1 - 4 434 3905169
BC093060.1 - 5 594 30409772
BC107703.1 - 4 437 6605146
BG472687.1 - 3 405

Gene Symbol
UQCRHL
Entrez Gene ID
440567
Gene Name
ubiquinol-cytochrome c reductase hinge protein like
Gene Aliases
Chromosome Location
Chr. 1: 15807162 - 15807699 on Build GRCh38
UniGene ID
Hs.568229
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001089591.1 NP_001083060.1 1 427

View Full Product Details