Gene Transcripts

Gene Symbol
C8orf58
Entrez Gene ID
541565
Gene Name
chromosome 8 open reading frame 58
Gene Aliases
Chromosome Location
Chr. 8: 22599601 - 22604150 on Build GRCh38
UniGene ID
Hs.743508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013842.2 NP_001013864.1 2 265
NM_001198827.1 NP_001185756.1 2 265
NM_173686.2 NP_775957.2 2 265
GenBank mRNA AK092034.1 - 1 73
AK295405.1 - 2 265
BC012750.2 - 1 56 3618901
BI910955.1 - 2 257
DC328974.1 - 2 265

View Full Product Details