Gene Transcripts

Gene Symbol
KRTAP19-8
Entrez Gene ID
728299
Gene Name
keratin associated protein 19-8
Gene Aliases
Chromosome Location
Chr. 21: 31038159 - 31038476 on Build GRCh38
UniGene ID
Hs.580875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099219.1 NP_001092689.1 1 181
GenBank mRNA AB096964.1 - 1 181

View Full Product Details