Gene Transcripts

Gene Symbol
Ms4a1
Entrez Gene ID
12482
Gene Name
membrane-spanning 4-domains, subfamily A, member 1
Gene Aliases
AA960661, Cd20, Ly-44, Ms4a2
Chromosome Location
Chr. 19: 11250603 - 11266151 on Build GRCm38
UniGene ID
Mm.4046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007641.5 NP_031667.1 6 746
GenBank mRNA AK017903.1 - 6 747
AK041428.1 - 6 745
AK080040.1 - 6 712
AK080148.1 - 744
AK137469.1 - 5 1909
AK156033.1 - 745
AK166571.1 - 6 840
AK166659.1 - 6 840
AK171631.1 - 6 775
AK171638.1 - 6 698
AK171659.1 - 6 731
BC028322.1 - 6 727 4503841
CT010253.1 - 5 627
M62541.1 - 804

View Full Product Details