Gene Transcripts

Gene Symbol
Cdr2
Entrez Gene ID
12585
Gene Name
cerebellar degeneration-related 2
Gene Aliases
AA617262
Chromosome Location
Chr. 7: 120957036 - 120982312 on Build GRCm38
UniGene ID
Mm.1640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007672.2 NP_031698.2 3, 4 606
GenBank mRNA AK148057.1 - 3, 4 590
AK171706.1 - 3, 4 561
BC117530.1 - 3, 4 495 40106297
BC118511.1 - 3, 4 495 40106298
U88588.1 - 4, 5 499

View Full Product Details