Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010259.2 | NP_034389.2 | 9, 10 | 1554 | |
GenBank mRNA | AK077796.1 | - | 9, 10 | 1556 | |
BC108990.2 | - | 1599 | 40047769 | ||
EF494422.1 | - | 9, 10 | 1554 | ||
M55544.1 | - | 1538 | |||
M63961.1 | - | 1632 |