Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010259.2 | NP_034389.2 | 10 | 1689 | |
GenBank mRNA | AK077796.1 | - | 10 | 1691 | |
BC108990.2 | - | 1734 | 40047769 | ||
EF494422.1 | - | 10 | 1689 | ||
M55544.1 | - | 1673 | |||
M63961.1 | - | 1767 |