Gene Transcripts

Gene Symbol
Hoxb7
Entrez Gene ID
15415
Gene Name
homeobox B7
Gene Aliases
AI325018, Hox-2.3
Chromosome Location
Chr. 11: 96286646 - 96290163 on Build GRCm38
UniGene ID
Mm.390366
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010460.2 NP_034590.2 1 331
GenBank mRNA AK136706.1 - 1 331
X06762.1 - 2413

View Full Product Details