Gene Transcripts

Gene Symbol
Magoh
Entrez Gene ID
17149
Gene Name
mago-nashi homolog, proliferation-associated (Drosophila)
Gene Aliases
Mago-m, Mos2
Chromosome Location
Chr. 4: 107879755 - 107887424 on Build GRCm38
UniGene ID
Mm.808
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282737.1 NP_001269666.1 3, 4 365
GenBank mRNA AF007862.1 - 3, 4 352
AF035939.1 - 3, 4 312
AK011512.1 - 3, 4 304
BC018176.1 - 3, 4 303 3587774
BY134232.1 - 3, 4 366

View Full Product Details