Gene Transcripts
- Gene Symbol
- Pip5k1a
- Entrez Gene ID
- 18720
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase, type 1 alpha
- Gene Aliases
- PI4P5K-I[a], PIP5K1-alpha, PIP5KIalpha, Pipk5a
- Chromosome Location
- Chr. 3: 95058530 - 95106930 on Build GRCm38
- UniGene ID
- Mm.296409
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293707.1 |
NP_001280636.1
|
6, 7 |
1073 |
|
NM_008847.3 |
NP_032873.2
|
6, 7 |
1070 |
|
GenBank mRNA |
AK098097.1 |
- |
6, 7 |
1027 |
|
AK150898.1 |
- |
6, 7 |
1030 |
|
AK151984.1 |
- |
6, 7 |
1030 |
|
AK158062.1 |
- |
6, 7 |
1015 |
|
AK196470.1 |
- |
2, 3 |
168 |
|
BC003763.1 |
- |
6, 7 |
709 |
3591003 |
BC031774.1 |
- |
6, 7 |
998 |
4503697 |
D86177.1 |
- |
6, 7 |
860 |
|
View Full Product Details