Gene Transcripts
- Gene Symbol
- Rtn3
- Entrez Gene ID
- 20168
- Gene Name
- reticulon 3
- Gene Aliases
- RTN3-A1
- Chromosome Location
- Chr. 19: 7425895 - 7483291 on Build GRCm38
- UniGene ID
- Mm.246990
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003933.2 |
NP_001003933.1
|
7, 8 |
2993 |
|
NM_001003934.2 |
NP_001003934.1
|
8, 9 |
3050 |
|
NM_001271486.1 |
NP_001258415.1
|
8, 9 |
2087 |
|
NM_001271487.1 |
NP_001258416.1
|
7, 8 |
926 |
|
NM_053076.3 |
NP_444306.1
|
6, 7 |
869 |
|
GenBank mRNA |
AB046114.1 |
- |
6, 7 |
863 |
|
AF195940.1 |
- |
6, 7 |
813 |
|
AK032109.1 |
- |
8, 9 |
2064 |
|
AK036892.1 |
- |
6, 7 |
835 |
|
AK049845.1 |
- |
6, 7 |
801 |
|
AK075883.1 |
- |
6, 7 |
798 |
|
AK088670.1 |
- |
6, 7 |
801 |
|
AK146505.1 |
- |
6, 7 |
677 |
|
AK148792.1 |
- |
6, 7 |
828 |
|
AK148947.1 |
- |
6, 7 |
830 |
|
AK165683.1 |
- |
6, 7 |
800 |
|
AY164700.1 |
- |
6, 7 |
863 |
|
AY427822.1 |
- |
7, 8 |
2974 |
|
AY507126.1 |
- |
6, 7 |
661 |
|
AY507127.1 |
- |
6, 7 |
2596 |
|
AY750849.1 |
- |
8, 9 |
3031 |
|
BC005478.1 |
- |
6, 7 |
772 |
|
BC014697.1 |
- |
6, 7 |
780 |
|
BC036717.1 |
- |
6, 7 |
778 |
|
BC079882.1 |
- |
8, 9 |
2960 |
6839323 |
View Full Product Details